Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170765.1 | NP_001164236.1 | 8 | 1520 | ||
NM_001170766.1 | NP_001164237.1 | 8 | 1520 | |||
NM_032440.3 | NP_115816.2 | 8 | 1540 | |||
XM_006718033.3 | XP_006718096.1 | 8 | 1828 | |||
XM_017016784.1 | XP_016872273.1 | 7 | 1451 | |||
XM_017016785.1 | XP_016872274.1 | 9 | 1595 | |||
XM_017016786.1 | XP_016872275.1 | 8 | 1516 | |||
XM_017016787.1 | XP_016872276.1 | 7 | 1388 | |||
XM_017016788.1 | XP_016872277.1 | 7 | 1442 | |||
GenBank mRNA | AB058698.1 | - | 8 | 1453 | ||
AL832044.1 | - | 1 | 279 | |||
AL832106.1 | - | 8 | 1500 | |||
AL834245.2 | - | 8 | 1523 | |||
BC053359.1 | - | 8 | 1461 | 6009825 |