Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170765.1 | NP_001164236.1 | 8 | 4774 | ||
NM_001170766.1 | NP_001164237.1 | 9 | 2197 | |||
NM_032440.3 | NP_115816.2 | 8 | 4794 | |||
XM_006718033.3 | XP_006718096.1 | 8 | 5082 | |||
XM_011540277.2 | XP_011538579.1 | 8 | 1623 | |||
XM_017016784.1 | XP_016872273.1 | 7 | 4705 | |||
XM_017016785.1 | XP_016872274.1 | 9 | 4849 | |||
XM_017016786.1 | XP_016872275.1 | 8 | 4770 | |||
XM_017016787.1 | XP_016872276.1 | 7 | 4642 | |||
XM_017016788.1 | XP_016872277.1 | 7 | 4696 | |||
GenBank mRNA | AB058698.1 | - | 8 | 4707 | ||
AK023248.1 | - | 1 | 2213 | |||
AK095345.1 | - | 1 | 3138 | |||
AL832044.1 | - | 1 | 3532 | |||
AL832106.1 | - | 8 | 4755 | |||
AL834245.2 | - | 8 | 4777 | |||
BC053359.1 | - | 9 | 2138 | 6009825 |