Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079692.2 | NP_001073160.1 | 4 | 1183 | ||
NM_001261827.1 | NP_001248756.1 | 5 | 1282 | |||
NM_001261828.1 | NP_001248757.1 | 6 | 1333 | |||
NM_032597.4 | NP_115986.3 | 5 | 1234 | |||
NR_049731.1 | 6 | 1403 | ||||
NR_049732.1 | 6 | 1355 | ||||
NR_049733.1 | 5 | 1304 | ||||
NR_049734.1 | 7 | 1454 | ||||
GenBank mRNA | AF367473.2 | - | 5 | 745 | ||
AK057418.1 | - | 6 | 949 | |||
AK314674.1 | - | 5 | 944 | |||
AL137391.1 | - | 1 | 101 | |||
AY094610.1 | - | 5 | 818 | |||
AY094611.1 | - | 4 | 767 | |||
AY584610.1 | - | 4 | 767 | |||
BC107431.1 | - | 6 | 860 |