Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSRC1 SIRNA Assays ›
Gene Symbol:
PSRC1
Gene Name:
proline and serine rich coiled-coil 1
Gene Aliases:
DDA3, FP3214
Chromosome Location:
Chr.1: 109279554 - 109283182 on Build GRCh38
UniGene ID:
Hs.405925
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
123700
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PSRC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005290.3
NP_001005290.1
8
1066
NM_001032291.2
NP_001027462.1
8
1172
NM_032636.7
NP_116025.1
8
1167
XM_005271283.2
XP_005271340.1
8
1166
XM_011542306.2
XP_011540608.1
7
1426
XM_017002560.1
XP_016858049.1
7
1267
XM_017002561.1
XP_016858050.1
7
1261
XM_017002562.1
XP_016858051.1
7
1291
XM_017002563.1
XP_016858052.1
7
1334
XM_017002564.1
XP_016858053.1
7
1265
XM_017002565.1
XP_016858054.1
7
1257
XM_017002566.1
XP_016858055.1
7
1282
XM_017002567.1
XP_016858056.1
7
1277
XM_017002568.1
XP_016858057.1
7
1474
XM_017002569.1
XP_016858058.1
8
1179
XM_017002570.1
XP_016858059.1
8
1183
XM_017002571.1
XP_016858060.1
8
1129
XM_017002572.1
XP_016858061.1
8
1123
XM_017002573.1
XP_016858062.1
8
1153
XM_017002574.1
XP_016858063.1
8
1126
XM_017002575.1
XP_016858064.1
8
1136
XM_017002576.1
XP_016858065.1
8
1075
XM_017002577.1
XP_016858066.1
7
1373
GenBank mRNA
AF223000.1
-
8
1146
AF447874.1
-
5
1310
AK126567.1
-
8
1002
AK289593.1
-
8
1109
BC056909.1
-
7
1275
6163125
BX649139.1
-
8
1013
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule bundle formation
mitotic metaphase plate congression
negative regulation of cell growth
positive regulation of microtubule polymerization
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of transcription, DNA-templated
cell division
regulation of mitotic spindle organization
protein binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0