Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144827.1 | NP_001138299.1 | 5, 6 | 755 | ||
NM_032795.2 | NP_116184.2 | 5, 6 | 848 | |||
XM_011543039.2 | XP_011541341.1 | 5, 6 | 845 | |||
GenBank mRNA | AK027400.1 | - | 5, 6 | 802 | ||
AK225320.1 | - | 5, 6 | 808 | |||
AK300723.1 | - | 3, 4 | 322 | |||
AK308059.1 | - | 5, 6 | 708 | |||
BC014131.2 | - | 5, 6 | 809 | 4621434 |