Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144827.1 | NP_001138299.1 | 7 | 1482 | ||
NM_032795.2 | NP_116184.2 | 7 | 1575 | |||
XM_011543039.2 | XP_011541341.1 | 7 | 1572 | |||
GenBank mRNA | AK027400.1 | - | 7 | 1529 | ||
AK300723.1 | - | 5 | 1049 | |||
AK308059.1 | - | 7 | 1435 | |||
BC014131.2 | - | 7 | 1536 | 4621434 |