Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286377.1 | NP_001273306.1 | 3 | 624 | ||
NM_001286378.1 | NP_001273307.1 | 5 | 490 | |||
NM_032858.2 | NP_116247.1 | 4 | 717 | |||
XM_006711583.1 | XP_006711646.1 | 5 | 521 | |||
XM_011510067.1 | XP_011508369.1 | 4 | 683 | |||
XM_011510068.1 | XP_011508370.1 | 5 | 474 | |||
XM_017002602.1 | XP_016858091.1 | 6 | 858 | |||
XM_017002603.1 | XP_016858092.1 | 5 | 520 | |||
GenBank mRNA | AK027810.1 | - | 4 | 524 | ||
AK131066.1 | - | 4 | 527 | |||
AK302253.1 | - | 3 | 440 | |||
AL133073.1 | - | 3 | 281 | |||
BC028595.2 | - | 4 | 497 | 5270252 | ||
BC034310.1 | - | 3 | 1002 | |||
DQ076156.2 | - | 4 | 526 | |||
GU383113.1 | - | 3 | 433 | |||
GU383114.1 | - | 5 | 490 |