Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286377.1 | NP_001273306.1 | 9 | 1185 | ||
NM_001286378.1 | NP_001273307.1 | 11 | 1051 | |||
NM_032858.2 | NP_116247.1 | 10 | 1278 | |||
XM_006711583.1 | XP_006711646.1 | 11 | 1082 | |||
XM_011510067.1 | XP_011508369.1 | 10 | 1244 | |||
XM_011510068.1 | XP_011508370.1 | 11 | 1035 | |||
XM_017002602.1 | XP_016858091.1 | 12 | 1419 | |||
XM_017002603.1 | XP_016858092.1 | 11 | 1081 | |||
GenBank mRNA | AK027810.1 | - | 10 | 1085 | ||
AK131066.1 | - | 10 | 1088 | |||
AK302253.1 | - | 9 | 1001 | |||
AL133073.1 | - | 9 | 842 | |||
BC028595.2 | - | 10 | 1058 | 5270252 | ||
BC034310.1 | - | 10 | 1706 | |||
DQ076156.2 | - | 10 | 1087 | |||
GU383113.1 | - | 9 | 994 | |||
GU383114.1 | - | 11 | 1051 |