Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134734.1 | NP_001128206.1 | 4 | 2264 | ||
NM_032884.4 | NP_116273.2 | 4 | 1395 | |||
GenBank mRNA | AK123355.1 | - | 4 | 1395 | ||
BC007637.1 | - | 4 | 1163 | 3529463 | ||
BC064845.1 | - | 4 | 1546 | 30390219 |