Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002901.2 | NP_002892.1 | 5, 6 | 1162 | ||
GenBank mRNA | AK094360.1 | - | 4, 5 | 1252 | ||
AK126419.1 | - | 1, 2 | 1493 | |||
AK129791.1 | - | 5, 6 | 1189 | |||
AY423728.1 | - | 3, 4 | 398 | |||
BC010120.2 | - | 5, 6 | 1031 | 3836263 | ||
D42073.1 | - | 5, 6 | 948 | |||
FJ224346.1 | - | 5, 6 | 948 |