Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033102.2 | NP_149093.1 | 5 | 2324 | ||
XM_005245556.3 | XP_005245613.1 | 5 | 2490 | |||
XM_005245557.4 | XP_005245614.1 | 5 | 2416 | |||
XM_017002638.1 | XP_016858127.1 | 5 | 2863 | |||
GenBank mRNA | AL832933.1 | - | 3 | 1388 | ||
AY033593.1 | - | 5 | 2268 | |||
BC050416.2 | - | 5 | 2232 | 6198823 |