Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 7 | 798 | ||
XM_011545352.2 | XP_011543654.1 | 8 | 967 | |||
XM_011545353.2 | XP_011543655.1 | 7 | 976 | |||
XM_017018560.1 | XP_016874049.1 | 7 | 831 | |||
GenBank mRNA | AK290866.1 | - | 7 | 794 | ||
AL834171.1 | - | 7 | 741 | |||
BC008013.1 | - | 3 | 251 | |||
BC009995.2 | - | 7 | 793 | 4129693 | ||
BC015664.2 | - | 6 | 691 | 4554197 |