Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 19, 20 | 2324 | ||
XM_011545352.2 | XP_011543654.1 | 20, 21 | 2493 | |||
XM_017018560.1 | XP_016874049.1 | 19, 20 | 2357 | |||
GenBank mRNA | AF172822.1 | - | 6, 7 | 710 | ||
AK290866.1 | - | 19, 20 | 2320 | |||
AL834171.1 | - | 19, 20 | 2268 | |||
BC006369.1 | - | 7, 8 | 693 | |||
BC008013.1 | - | 15, 16 | 1777 | |||
BC009859.2 | - | 9, 10 | 983 | |||
BC015664.2 | - | 20, 21 | 2634 | 4554197 |
Gene Symbol: INTS4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027392.2 | 11, 12 | 1468 |
Gene Symbol: LOC107986705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012880.1 | XP_016868369.1 | 4, 5 | 896 | ||
XM_017012881.1 | XP_016868370.1 | 4, 5 | 354 | |||
XM_017012882.1 | XP_016868371.1 | 2, 3 | 393 |