Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159397.1 | NP_001152869.1 | 10 | 1887 | ||
NM_001159398.1 | NP_001152870.1 | 11 | 2011 | |||
NM_052938.4 | NP_443170.1 | 11 | 2014 | |||
XM_005244866.2 | XP_005244923.1 | 11 | 1975 | |||
XM_005244867.2 | XP_005244924.1 | 10 | 1868 | |||
XM_005244869.3 | XP_005244926.1 | 9 | 1710 | |||
XM_011509125.1 | XP_011507427.1 | 12 | 1983 | |||
XM_011509126.1 | XP_011507428.1 | 12 | 1980 | |||
XM_011509127.2 | XP_011507429.1 | 10 | 1977 | |||
XM_011509128.1 | XP_011507430.1 | 11 | 1987 | |||
XM_011509129.1 | XP_011507431.1 | 11 | 1984 | |||
XM_011509130.1 | XP_011507432.1 | 11 | 1972 | |||
XM_011509133.2 | XP_011507435.1 | 11 | 1839 | |||
XM_011509134.2 | XP_011507436.1 | 10 | 1814 | |||
XM_011509135.2 | XP_011507437.1 | 10 | 1731 | |||
XM_011509137.2 | XP_011507439.1 | 8 | 1359 | |||
XM_017000227.1 | XP_016855716.1 | 10 | 3981 | |||
GenBank mRNA | AF459634.1 | - | 11 | 1967 | ||
AK096690.1 | - | 10 | 1826 | |||
AL833674.1 | - | 7 | 1058 | |||
AL833970.1 | - | 7 | 2002 | |||
AY043464.1 | - | 11 | 1969 | |||
BC021274.2 | - | 11 | 1682 | |||
BC033690.1 | - | 11 | 1965 | 5211491 |