Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052939.3 | NP_443171.2 | 15 | 2479 | ||
NR_135214.1 | 15 | 2512 | ||||
NR_135215.1 | 15 | 2494 | ||||
NR_135216.1 | 15 | 2399 | ||||
NR_135217.1 | 14 | 2209 | ||||
XM_006711145.1 | XP_006711208.1 | 15 | 2409 | |||
GenBank mRNA | AF416901.1 | - | 15 | 2358 | ||
AF416902.1 | - | 14 | 2073 | |||
AF416903.1 | - | 15 | 2376 | |||
AF416904.1 | - | 14 | 2592 | |||
AF416905.1 | - | 15 | 2263 | |||
AF459027.1 | - | 15 | 2479 | |||
AK125622.1 | - | 4 | 312 | |||
AK301257.1 | - | 14 | 2615 | |||
AL833391.1 | - | 9 | 3029 | |||
AY043466.1 | - | 14 | 2283 |