Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256409.1 | NP_001243338.1 | 6 | 1028 | ||
NM_052940.4 | NP_443172.1 | 5 | 994 | |||
XM_006710327.3 | XP_006710390.1 | 6 | 850 | |||
XM_006710328.3 | XP_006710391.1 | 6 | 1363 | |||
GenBank mRNA | AK074521.1 | - | 5 | 967 | ||
AK075201.1 | - | 5 | 956 | |||
AK090881.1 | - | 6 | 1277 | |||
AK095767.1 | - | 6 | 947 | |||
BC013101.1 | - | 5 | 833 | 3918785 |