Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM46B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052943.3 | NP_443175.2 | 2 | 1664 | ||
GenBank mRNA | AK122816.1 | - | 2 | 1794 | ||
BC012790.2 | - | 2 | 1658 | 3957388 | ||
BC014160.2 | - | 2 | 1658 | 4542896 |