Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053282.4 | NP_444512.2 | 4 | 1654 | ||
GenBank mRNA | AF484964.1 | - | 4 | 1631 | ||
AK097162.1 | - | 4 | 1581 | |||
BC022407.1 | - | 4 | 1642 | 4717041 | ||
BC066595.1 | - | 3 | 1463 | 5755414 |