Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRASLS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146728.1 | NP_001140200.1 | 6 | 958 | ||
NM_001146729.1 | NP_001140201.1 | 6 | 978 | |||
NM_054108.3 | NP_473449.1 | 6 | 1008 | |||
XM_011544751.2 | XP_011543053.1 | 6 | 913 | |||
XM_011544752.2 | XP_011543054.1 | 3 | 613 | |||
XM_011544753.1 | XP_011543055.1 | 7 | 832 | |||
XM_017017172.1 | XP_016872661.1 | 7 | 820 | |||
GenBank mRNA | AJ416558.1 | - | 6 | 995 | ||
AK055392.1 | - | 4 | 494 | |||
AK302312.1 | - | 6 | 919 | |||
BC034222.1 | - | 6 | 937 | 5268703 |