Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138300.3 | NP_612157.1 | 3 | 2280 | ||
GenBank mRNA | AF289598.1 | - | 3 | 2031 | ||
AK090545.1 | - | 1 | 1598 | |||
AK092389.1 | - | 3 | 2229 | |||
AK095425.1 | - | 1 | 1039 | |||
BC006132.1 | - | 3 | 2246 | 3627860 | ||
BC013725.1 | - | 1 | 1556 | |||
BC032099.2 | - | 3 | 2246 | 4126360 |