Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080407.2 | NP_001073876.2 | 2 | 450 | ||
XM_011542566.2 | XP_011540868.1 | 1 | 520 | |||
XM_011542567.2 | XP_011540869.1 | 1 | 520 | |||
XM_011542568.2 | XP_011540870.1 | 1 | 520 | |||
XM_011542569.2 | XP_011540871.1 | 1 | 520 | |||
XM_011542570.2 | XP_011540872.1 | 1 | 520 | |||
XM_011542571.2 | XP_011540873.1 | 1 | 520 | |||
XM_017017155.1 | XP_016872644.1 | 2 | 1384 | |||
XM_017017156.1 | XP_016872645.1 | 2 | 1384 | |||
GenBank mRNA | BC011001.2 | - | 2 | 450 |