Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164824.1 | NP_001158296.1 | 3 | 1308 | ||
NM_001164825.1 | NP_001158297.1 | 2 | 1201 | |||
NM_001320261.1 | NP_001307190.1 | 2 | 1083 | |||
NM_138428.5 | NP_612437.3 | 2 | 1058 | |||
XM_005270403.4 | XP_005270460.1 | 2 | 1541 | |||
XM_011540553.2 | XP_011538855.1 | 2 | 943 | |||
XM_011540554.2 | XP_011538856.2 | 2 | 2254 | |||
XM_011540555.2 | XP_011538857.1 | 2 | 1070 | |||
XM_017000166.1 | XP_016855655.1 | 2 | 1062 | |||
GenBank mRNA | AK074853.1 | - | 2 | 990 | ||
BC011880.1 | - | 2 | 984 | 3607345 | ||
BC034598.1 | - | 2 | 2799 | |||
BC064320.1 | - | 2 | 1021 | |||
BC073851.1 | - | 2 | 984 | 5499672 | ||
BC107421.1 | - | 2 | 983 | 4688408 |