Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138440.2 | NP_612449.2 | 2 | 2173 | ||
GenBank mRNA | AY358298.1 | - | 3 | 1929 | ||
AY358299.1 | - | 2 | 2137 | |||
BC013767.1 | - | 1 | 1802 | |||
BC068575.1 | - | 2 | 2173 | 30349094 |