Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138788.3 | NP_620143.1 | 2 | 179 | ||
XM_005271400.4 | XP_005271457.1 | 3 | 304 | |||
XM_011542586.2 | XP_011540888.1 | 3 | 348 | |||
XM_011542588.2 | XP_011540890.1 | 3 | 299 | |||
XM_017017188.1 | XP_016872677.1 | 4 | 454 | |||
XM_017017189.1 | XP_016872678.1 | 4 | 470 | |||
GenBank mRNA | AK098106.1 | - | 2 | 180 | ||
AK290283.1 | - | 2 | 175 | |||
AK292058.1 | - | 2 | 175 | |||
BC016153.1 | - | 2 | 558 | 3841102 |