Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YY1AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198899.1 | NP_001185828.1 | 8 | 1914 | ||
NM_001198900.1 | NP_001185829.1 | 10 | 1847 | |||
NM_001198901.1 | NP_001185830.1 | 10 | 1737 | |||
NM_001198902.1 | NP_001185831.1 | 10 | 1704 | |||
NM_001198903.1 | NP_001185832.1 | 10 | 2224 | |||
NM_001198904.1 | NP_001185833.1 | 10 | 2164 | |||
NM_001198905.1 | NP_001185834.1 | 11 | 1828 | |||
NM_001198906.1 | NP_001185835.1 | 11 | 1892 | |||
NM_018253.3 | NP_060723.2 | 12 | 1969 | |||
NM_139118.2 | NP_620829.1 | 11 | 1812 | |||
NM_139119.2 | NP_620830.1 | 11 | 1885 | |||
NM_139121.2 | NP_620832.1 | 11 | 1838 | |||
GenBank mRNA | AF203474.1 | - | 10 | 1844 | ||
AF206328.1 | - | 10 | 1659 | |||
AF466401.1 | - | 10 | 1678 | |||
AK001737.1 | - | 7 | 1403 | |||
AK023976.1 | - | 11 | 1780 | |||
AK125590.1 | - | 4 | 1661 | |||
AK225883.1 | - | 7 | 1454 | |||
AK297562.1 | - | 10 | 2037 | |||
AK303386.1 | - | 8 | 1892 | |||
AK303944.1 | - | 6 | 1271 | |||
AK315049.1 | - | 11 | 1859 | |||
AY604179.1 | - | 10 | 1709 | |||
BC001655.2 | - | 12 | 1899 | 2961648 | ||
BC001843.2 | - | 12 | 1899 | 2961648 | ||
BC003500.1 | - | 11 | 1838 | |||
BC008766.2 | - | 10 | 1721 | 3051391 | ||
BC009358.2 | - | 7 | 1465 | |||
BC014906.2 | - | 11 | 1817 | 3922542 | ||
CR749789.1 | - | 11 | 1855 |