Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144567.4 | NP_653168.2 | 2 | 402 | ||
XM_005273344.1 | XP_005273401.1 | 2 | 423 | |||
XM_005273345.1 | XP_005273402.1 | 2 | 469 | |||
XM_005273346.2 | XP_005273403.1 | 2 | 222 | |||
XM_017002774.1 | XP_016858263.1 | 2 | 351 | |||
XM_017002775.1 | XP_016858264.1 | 2 | 399 | |||
XM_017002776.1 | XP_016858265.1 | 2 | 423 | |||
XM_017002777.1 | XP_016858266.1 | 2 | 469 | |||
XM_017002778.1 | XP_016858267.1 | 2 | 212 | |||
GenBank mRNA | AK295116.1 | - | 2 | 331 | ||
AL079275.1 | - | 2 | 357 | |||
AY468376.1 | - | 1 | 132 | |||
BC016966.1 | - | 1 | 146 | |||
DA718585.1 | - | 2 | 423 | |||
DC395916.1 | - | 2 | 402 |