Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HENMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102592.1 | NP_001096062.1 | 5 | 547 | ||
NM_144584.2 | NP_653185.2 | 5 | 730 | |||
XM_005270411.1 | XP_005270468.1 | 4 | 429 | |||
XM_017000179.1 | XP_016855668.1 | 4 | 531 | |||
GenBank mRNA | AK055087.1 | - | 5 | 730 | ||
AK310152.1 | - | 5 | 720 | |||
BC012198.1 | - | 5 | 456 | 2906200 | ||
BC088366.1 | - | 5 | 467 | 30390911 | ||
BP378034.1 | - | 5 | 547 |