Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256385.1 | NP_001243314.1 | 7 | 840 | ||
NM_001256386.1 | NP_001243315.1 | 7 | 840 | |||
NM_001256387.1 | NP_001243316.1 | 6 | 783 | |||
NM_144620.3 | NP_653221.1 | 7 | 823 | |||
GenBank mRNA | AK096892.1 | - | 7 | 766 | ||
AL833417.1 | - | 7 | 788 | |||
AY358131.1 | - | 6 | 756 | |||
BC009613.1 | - | 7 | 765 | 4245944 |