Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144622.2 | NP_653223.2 | 4 | 700 | ||
XM_011509188.2 | XP_011507490.1 | 2 | 163 | |||
GenBank mRNA | AK057496.1 | - | 4 | 700 | ||
AK097468.1 | - | 2 | 114 | |||
BC111794.1 | - | 4 | 679 |