Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144666.2 | NP_653267.2 | 43 | 14740 | ||
GenBank mRNA | AB290171.1 | - | 30 | 11440 | ||
AK057314.1 | - | 10 | 3259 | |||
AK128064.1 | - | 11 | 4382 | |||
AL137690.1 | - | 6 | 2704 | |||
AL834222.1 | - | 9 | 4346 | |||
AY358770.1 | - | 7 | 1653 | |||
BC082757.1 | - | 9 | 4195 | |||
BC117301.1 | - | 10 | 3084 | |||
BC117303.1 | - | 10 | 3084 | |||
BC143910.1 | - | 10 | 3054 |