Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286834.1 | NP_001273763.1 | 7 | 789 | ||
NM_001286837.1 | NP_001273766.1 | 7 | 1027 | |||
NM_002936.4 | NP_002927.2 | 7 | 789 | |||
GenBank mRNA | AF039652.1 | - | 7 | 759 | ||
AF048994.1 | - | 7 | 705 | |||
AF048995.1 | - | 7 | 739 | |||
AK075490.1 | - | 7 | 738 | |||
AK096913.1 | - | 7 | 945 | |||
BC002973.1 | - | 7 | 748 | 3537074 | ||
BT019670.1 | - | 7 | 678 | |||
CR541820.1 | - | 7 | 678 |