Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308339.1 | NP_001295268.1 | 5 | 517 | ||
NM_144973.3 | NP_659410.3 | 3 | 447 | |||
XM_005253326.2 | XP_005253383.1 | 4 | 449 | |||
XM_005253327.3 | XP_005253384.1 | 3 | 442 | |||
XM_005253330.3 | XP_005253387.1 | 4 | 449 | |||
XM_006719040.3 | XP_006719103.1 | 4 | 449 | |||
XM_017018896.1 | XP_016874385.1 | 4 | 449 | |||
XM_017018897.1 | XP_016874386.1 | 4 | 812 | |||
XM_017018898.1 | XP_016874387.1 | 4 | 449 | |||
XM_017018899.1 | XP_016874388.1 | 4 | 339 | |||
GenBank mRNA | AI762929.1 | - | 3 | 446 | ||
AK125323.1 | - | 5 | 517 | |||
BC020855.1 | - | 2 | 117 | |||
BC068580.1 | - | 4 | 504 | 5265807 |