Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308339.1 | NP_001295268.1 | 23 | 6070 | ||
NM_144973.3 | NP_659410.3 | 21 | 6000 | |||
XM_005253326.2 | XP_005253383.1 | 23 | 6113 | |||
XM_005253327.3 | XP_005253384.1 | 22 | 6106 | |||
XM_005253330.3 | XP_005253387.1 | 23 | 5582 | |||
XM_006719040.3 | XP_006719103.1 | 22 | 6002 | |||
XM_011520590.2 | XP_011518892.1 | 20 | 5349 | |||
XM_017018896.1 | XP_016874385.1 | 21 | 5903 | |||
XM_017018897.1 | XP_016874386.1 | 23 | 6476 | |||
XM_017018898.1 | XP_016874387.1 | 22 | 5576 | |||
XM_017018899.1 | XP_016874388.1 | 22 | 5361 | |||
GenBank mRNA | AB209341.1 | - | 17 | 4631 | ||
AK021465.1 | - | 1 | 713 | |||
AK123642.1 | - | 4 | 1628 | |||
BX537924.1 | - | 14 | 3773 | |||
CR749639.1 | - | 9 | 3242 |