Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168247.2 | NP_001161719.1 | 6 | 1926 | ||
NM_001300859.1 | NP_001287788.1 | 6 | 1656 | |||
NM_144990.3 | NP_659427.3 | 6 | 1770 | |||
XM_005270597.2 | XP_005270654.1 | 5 | 2509 | |||
XM_005270598.2 | XP_005270655.1 | 5 | 1825 | |||
XM_005270599.2 | XP_005270656.1 | 6 | 1833 | |||
XM_006710432.3 | XP_006710495.1 | 5 | 2920 | |||
XM_011540943.1 | XP_011539245.1 | 5 | 2456 | |||
XM_011540945.1 | XP_011539247.1 | 5 | 2393 | |||
XM_011540947.1 | XP_011539249.1 | 5 | 1918 | |||
XM_011540948.1 | XP_011539250.1 | 6 | 2371 | |||
XM_011540949.1 | XP_011539251.1 | 5 | 2301 | |||
XM_011540950.1 | XP_011539252.1 | 5 | 2271 | |||
XM_011540951.1 | XP_011539253.1 | 6 | 1825 | |||
XM_011540952.2 | XP_011539254.1 | 5 | 2248 | |||
XM_011540953.2 | XP_011539255.1 | 5 | 2828 | |||
XM_011540954.1 | XP_011539256.1 | 5 | 2179 | |||
XM_017000573.1 | XP_016856062.1 | 4 | 2365 | |||
XM_017000574.1 | XP_016856063.1 | 5 | 2329 | |||
GenBank mRNA | AK097609.1 | - | 6 | 1644 | ||
BC022037.2 | - | 6 | 1732 | 4826722 | ||
BC037879.1 | - | 3 | 2438 | 5274032 | ||
BC050353.1 | - | 4 | 4090 | 5265392 |