Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145018.3 | NP_659455.3 | 6 | 670 | ||
XM_011544835.1 | XP_011543137.1 | 5 | 532 | |||
XM_011544836.2 | XP_011543138.1 | 5 | 487 | |||
XM_011544837.1 | XP_011543139.1 | 4 | 382 | |||
XM_011544838.2 | XP_011543140.1 | 5 | 516 | |||
XM_017017352.1 | XP_016872841.1 | 4 | 387 | |||
XM_017017353.1 | XP_016872842.1 | 4 | 354 | |||
GenBank mRNA | AK058145.1 | - | 7 | 881 | ||
AK096157.1 | - | 6 | 635 | |||
AK292399.1 | - | 6 | 629 | |||
AK297366.1 | - | 4 | 355 | |||
AK299581.1 | - | 5 | 510 | |||
BC039268.1 | - | 6 | 638 | 5296465 |