Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288563.1 | NP_001275492.1 | 8 | 1098 | ||
NM_145172.4 | NP_660155.2 | 9 | 1215 | |||
GenBank mRNA | AK054629.1 | - | 8 | 1052 | ||
AK292603.1 | - | 9 | 1186 | |||
AY049724.1 | - | 9 | 1196 | |||
BC040265.1 | - | 9 | 1169 | 5165779 | ||
BX648851.1 | - | 9 | 1221 |