Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRTOMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205138.3 | NP_001192067.1 | 5 | 2187 | ||
NM_001271471.2 | NP_001258400.1 | 5 | 2606 | |||
NM_001318803.1 | NP_001305732.1 | 6 | 2281 | |||
NM_145309.5 | NP_660352.1 | 6 | 2324 | |||
NR_026886.3 | 7 | 2504 | ||||
NR_134858.1 | 4 | 2103 | ||||
XM_006718473.3 | XP_006718536.1 | 6 | 2550 | |||
XM_006718474.3 | XP_006718537.1 | 5 | 2589 | |||
XM_011544847.2 | XP_011543149.1 | 6 | 2778 | |||
XM_011544848.2 | XP_011543150.1 | 7 | 2836 | |||
XM_017017359.1 | XP_016872848.1 | 7 | 2941 | |||
XM_017017360.1 | XP_016872849.1 | 7 | 2793 | |||
GenBank mRNA | BC012855.1 | - | 6 | 1796 | 3863323 | |
EU627066.1 | - | 6 | 1796 | |||
EU627068.1 | - | 5 | 2077 |