Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320252.1 | NP_001307181.1 | 7 | 2449 | ||
NM_152281.2 | NP_689494.2 | 5 | 2149 | |||
NR_027397.1 | 5 | 2153 | ||||
XM_006711628.3 | XP_006711691.1 | 7 | 2424 | |||
XM_011510149.2 | XP_011508451.1 | 5 | 2086 | |||
XM_011510150.2 | XP_011508452.1 | 6 | 2327 | |||
XM_011510151.1 | XP_011508453.1 | 5 | 2076 | |||
XM_017002807.1 | XP_016858296.1 | 7 | 2390 | |||
GenBank mRNA | AK021814.1 | - | 5 | 2142 | ||
AK057661.1 | - | 5 | 1959 | |||
BC047476.1 | - | 7 | 2420 | 5296051 |