Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152315.3 | NP_689528.2 | 6 | 1444 | ||
XM_011542594.2 | XP_011540896.1 | 8 | 1961 | |||
XM_011542595.2 | XP_011540897.1 | 9 | 2073 | |||
XM_011542596.2 | XP_011540898.1 | 9 | 2314 | |||
XM_011542597.2 | XP_011540899.1 | 8 | 2202 | |||
XM_011542598.2 | XP_011540900.1 | 7 | 1902 | |||
XM_011542599.2 | XP_011540901.1 | 7 | 2143 | |||
GenBank mRNA | BC029049.1 | - | 6 | 1407 | 5186472 | |
BX648601.1 | - | 4 | 1807 |