Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL14EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152316.2 | NP_689529.1 | 4 | 1506 | ||
XM_005252792.3 | XP_005252849.1 | 4 | 1957 | |||
GenBank mRNA | AK096287.1 | - | 4 | 1503 | ||
BX647617.1 | - | 4 | 1500 |