Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4K4 SIRNA Assays ›
Gene Symbol:
MAP4K4
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases:
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location:
Chr.2: 101697703 - 101894690 on Build GRCh38
UniGene ID:
Hs.701013
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1240
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4K4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242559.1
NP_001229488.1
2, 3
136
NM_001242560.1
NP_001229489.1
2, 3
509
NM_004834.4
NP_004825.3
2, 3
509
NM_145686.3
NP_663719.2
2, 3
509
NM_145687.3
NP_663720.1
2, 3
136
XM_005264048.2
XP_005264105.1
2, 3
205
XM_005264050.3
XP_005264107.1
2, 3
205
XM_005264052.3
XP_005264109.1
2, 3
205
XM_005264057.3
XP_005264114.1
2, 3
205
XM_005264058.3
XP_005264115.1
2, 3
205
XM_005264059.3
XP_005264116.1
2, 3
205
XM_005264060.3
XP_005264117.1
2, 3
205
XM_005264062.3
XP_005264119.1
2, 3
205
XM_005264063.3
XP_005264120.1
2, 3
205
XM_005264064.3
XP_005264121.1
2, 3
205
XM_005264065.3
XP_005264122.1
2, 3
205
XM_005264066.3
XP_005264123.1
2, 3
205
XM_005264068.3
XP_005264125.1
2, 3
205
XM_005264069.2
XP_005264126.1
2, 3
205
XM_005264071.2
XP_005264128.1
2, 3
205
XM_006712865.2
XP_006712928.1
2, 3
205
XM_006712868.2
XP_006712931.1
2, 3
205
XM_006712869.2
XP_006712932.1
2, 3
205
XM_017005349.1
XP_016860838.1
2, 3
205
XM_017005350.1
XP_016860839.1
2, 3
205
XM_017005351.1
XP_016860840.1
2, 3
205
XM_017005352.1
XP_016860841.1
2, 3
205
XM_017005353.1
XP_016860842.1
2, 3
205
XM_017005354.1
XP_016860843.1
2, 3
205
XM_017005355.1
XP_016860844.1
2, 3
205
XM_017005356.1
XP_016860845.1
2, 3
205
XM_017005357.1
XP_016860846.1
2, 3
205
XM_017005358.1
XP_016860847.1
2, 3
205
XM_017005359.1
XP_016860848.1
2, 3
205
XM_017005360.1
XP_016860849.1
2, 3
205
XM_017005361.1
XP_016860850.1
2, 3
205
XM_017005362.1
XP_016860851.1
2, 3
205
XM_017005363.1
XP_016860852.1
2, 3
205
XM_017005364.1
XP_016860853.1
2, 3
205
XM_017005365.1
XP_016860854.1
2, 3
205
XM_017005366.1
XP_016860855.1
2, 3
205
XM_017005367.1
XP_016860856.1
2, 3
205
XM_017005368.1
XP_016860857.1
2, 3
205
XM_017005369.1
XP_016860858.1
2, 3
205
XM_017005370.1
XP_016860859.1
2, 3
205
XM_017005371.1
XP_016860860.1
2, 3
205
XM_017005372.1
XP_016860861.1
2, 3
205
XM_017005373.1
XP_016860862.1
2, 3
205
XM_017005374.1
XP_016860863.1
2, 3
205
XM_017005375.1
XP_016860864.1
2, 3
205
GenBank mRNA
AB014587.1
-
1
20
AF096300.1
-
2, 3
210
AK296396.1
-
2, 3
205
AY212247.1
-
2, 3
131
AY834276.1
-
2, 3
131
DB086475.1
-
3, 4
352
DR002278.1
-
2, 3
518
EU794634.1
-
2, 3
186
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008696, NM_001252200, NM_001252201, NM_001252202, BC141109, and others.
Matching Taqman Gene Expression Assay:
Hs00377410_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction by protein phosphorylation
microvillus assembly
intracellular signal transduction
negative regulation of apoptotic process
regulation of JNK cascade
creatine kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline