Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199679.1 | NP_001186608.1 | 4 | 651 | ||
NM_152487.2 | NP_689700.1 | 4 | 566 | |||
GenBank mRNA | AK056404.1 | - | 4 | 476 | ||
BC137457.1 | - | 4 | 366 | 9021457 | ||
BC137458.1 | - | 4 | 366 | 9021458 | ||
BC139737.1 | - | 4 | 366 | 40053604 |
Gene Symbol: TMEM56-RWDD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199691.1 | NP_001186620.1 | 4 | 455 |