Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM20D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152491.4 | NP_689704.4 | 6 | 873 | ||
NR_135186.1 | 6 | 873 | ||||
XM_017000396.1 | XP_016855885.1 | 6 | 867 | |||
XM_017000397.1 | XP_016855886.1 | 6 | 867 | |||
XM_017000398.1 | XP_016855887.1 | 6 | 867 | |||
GenBank mRNA | AK057131.1 | - | 6 | 857 | ||
AK290786.1 | - | 6 | 857 | |||
BC039170.1 | - | 6 | 842 | 4746689 | ||
BC063477.1 | - | 6 | 842 | 5221187 |