Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152492.2 | NP_689705.2 | 12 | 1957 | ||
XM_011540780.1 | XP_011539082.1 | 12 | 2085 | |||
XM_017000402.1 | XP_016855891.1 | 11 | 2316 | |||
XM_017000403.1 | XP_016855892.1 | 11 | 2191 | |||
XM_017000406.1 | XP_016855895.1 | 12 | 2130 | |||
GenBank mRNA | AK057387.1 | - | 12 | 1960 | ||
BC112108.1 | - | 12 | 1958 | 8327576 | ||
BC112112.1 | - | 12 | 1957 | 8327580 |