Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143687.2 | NP_001137159.1 | 10 | 1247 | ||
NM_152494.3 | NP_689707.2 | 11 | 1322 | |||
GenBank mRNA | AK057347.1 | - | 11 | 1304 | ||
AK301876.1 | - | 10 | 1224 | |||
AK301902.1 | - | 11 | 1401 | |||
AK301935.1 | - | 10 | 1033 | |||
BC064844.1 | - | 10 | 1277 | 30378702 |