Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143687.2 | NP_001137159.1 | 16 | 2195 | ||
NM_152494.3 | NP_689707.2 | 17 | 2270 | |||
GenBank mRNA | AK057347.1 | - | 17 | 2252 | ||
AK301876.1 | - | 16 | 2172 | |||
AK301935.1 | - | 16 | 1981 |