Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152587.3 | NP_689800.3 | 5 | 441 | ||
XM_005271412.3 | XP_005271469.1 | 4 | 403 | |||
XM_005271413.3 | XP_005271470.1 | 4 | 364 | |||
XM_005271414.3 | XP_005271471.1 | 5 | 457 | |||
XM_005271415.4 | XP_005271472.1 | 5 | 457 | |||
XM_011542639.2 | XP_011540941.1 | 6 | 494 | |||
XM_011542640.2 | XP_011540942.1 | 5 | 457 | |||
XM_011542641.2 | XP_011540943.1 | 3 | 294 | |||
XM_011542642.1 | XP_011540944.1 | 5 | 457 | |||
XM_011542643.2 | XP_011540945.1 | 5 | 457 | |||
XM_017017246.1 | XP_016872735.1 | 5 | 401 | |||
XM_017017247.1 | XP_016872736.1 | 5 | 457 | |||
XM_017017248.1 | XP_016872737.1 | 5 | 457 | |||
XM_017017249.1 | XP_016872738.1 | 3 | 294 | |||
XM_017017250.1 | XP_016872739.1 | 4 | 364 | |||
GenBank mRNA | AK303096.1 | - | 3 | 294 | ||
BC029536.1 | - | 5 | 414 | 5298158 | ||
DB104722.1 | - | 3 | 294 |