Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152760.2 | NP_689973.2 | 4 | 435 | ||
XM_006718488.3 | XP_006718551.1 | 4 | 738 | |||
XM_017017470.1 | XP_016872959.1 | 4 | 713 | |||
XM_017017471.1 | XP_016872960.1 | 5 | 897 | |||
XM_017017472.1 | XP_016872961.1 | 3 | 1099 | |||
XM_017017473.1 | XP_016872962.1 | 3 | 1099 | |||
GenBank mRNA | AK055496.1 | - | 4 | 395 | ||
AK289587.1 | - | 4 | 401 | |||
AK293827.1 | - | 3 | 541 | |||
AK299512.1 | - | 4 | 346 | |||
AK309435.1 | - | 2 | 856 | |||
AK309789.1 | - | 3 | 492 | |||
AK311588.1 | - | 3 | 1016 | |||
AL833261.1 | - | 4 | 421 | |||
BC040981.2 | - | 4 | 411 | 4797976 | ||
BC045563.1 | - | 4 | 384 | 4815203 |