Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098526.1 | NP_001091996.1 | 5 | 658 | ||
NM_001286570.1 | NP_001273499.1 | 4 | 595 | |||
NM_001286571.1 | NP_001273500.1 | 4 | 886 | |||
NM_153206.2 | NP_694938.2 | 4 | 896 | |||
NR_104479.1 | 4 | 886 | ||||
XM_005271407.3 | XP_005271464.1 | 4 | 596 | |||
XM_017017213.1 | XP_016872702.1 | 5 | 583 | |||
GenBank mRNA | AJ515553.2 | - | 5 | 578 | ||
AK090409.1 | - | 5 | 558 | |||
AK094399.1 | - | 4 | 896 | |||
AK298722.1 | - | 4 | 512 | |||
AK301099.1 | - | 5 | 575 | |||
AY093686.1 | - | 5 | 575 | |||
AY093687.1 | - | 3 | 368 | |||
AY138965.1 | - | 5 | 559 | |||
AY358362.1 | - | 5 | 568 | |||
BC100794.2 | - | 4 | 585 | 40000649 | ||
BC100795.2 | - | 4 | 585 | 40000652 | ||
BC100796.1 | - | 4 | 585 | 40000654 | ||
BC100797.1 | - | 4 | 585 | 40000656 |