Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153367.3 | NP_699198.2 | 4 | 1278 | ||
XM_011539452.2 | XP_011537754.1 | 4 | 1229 | |||
GenBank mRNA | AK024784.1 | - | 1 | 21 | ||
AK075279.1 | - | 4 | 1130 | |||
AK293361.1 | - | 4 | 917 | |||
AK303734.1 | - | 4 | 909 | |||
BC028617.1 | - | 4 | 1280 | 4821347 | ||
BC039249.1 | - | 4 | 1272 | |||
BC053564.1 | - | 4 | 1177 | 6193781 | ||
BX648607.1 | - | 1 | 164 |